Nonadecylbenzene 98,0 GC_ANALYSMETOD
Leverantör: TCI
|
Fara
|
Synonyms:
1-Phenylnonadecane
N0349-25G
N0349-5G
TCIAN0349-25GEA
4750
SEK
TCIAN0349-25G
TCIAN0349-5G
Nonadecylbenzene 98,0 GC_ANALYSMETOD
Nonadecylbenzene
Formel:
C₂₅H₄₄ MW: 344.62 g/mol Kokpunkt: 419 Smältpunkt: 40 Storage Temperature: Omgivande |
MDL Number:
MFCD00026692 CAS nummer: 29136-19-4 EINECS: 249-467-4 |
|
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Freezing point | 23.0 to 30.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...