Dodecylbenzene ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
n-Dodecylbenzene, 1-Phenyldodecane
D1074-25ML
D1074-500ML
TCIAD1074-25MLEA
645
SEK
TCIAD1074-25ML
TCIAD1074-500ML
Dodecylbenzene ≥98.0%
Dodecylbenzene
Formel:
C₁₈H₃₀ MW: 246,44 g/mol Kokpunkt: 148°C Smältpunkt: –4 °C Densitet: 0,85 g/cm³ (20 °C) Flampunkt: 124°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008974 CAS nummer: 123-01-3 EINECS: 204-591-8 |
Specifikation testresultat
Appearance | Colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...