Isopentylbenzene ≥98.0%

Leverantör: TCI

Synonyms: Isoamylbenzene, (3-Methylbutyl)benzene

M0656-25ML M0656-5ML
TCIAM0656-25MLEA 1800 SEK
TCIAM0656-25ML TCIAM0656-5ML
Isopentylbenzene ≥98.0%
Isopentylbenzene
Formel: (C₆H₅)CH₂CH₂CH(CH₃)₂
MW: 148.25 g/mol
Kokpunkt: 199°C
Flampunkt: 65°C
Storage Temperature: Omgivande
MDL Number: MFCD00008948
CAS nummer: 2049-94-7
EINECS: 218-075-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69f971f3ee3bc24feb078f16e3df83b4d7555b58&vid=4043c320432ef9ebd62cf2e0a4d0e400c098d673&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 0.8540 to 0.8580
Refractive index n20/D 1.4840 to 1.4880

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR