4-Isopropylphenol ≥99.0%
Leverantör: TCI EUROPE
|
Fara
|
I0262-25G
I0262-500G
I0262-100G
TCIAI0262-25GEA
750
SEK
TCIAI0262-25G
TCIAI0262-500G
TCIAI0262-100G
4-Isopropylphenol ≥99.0%
4-Isopropylphenol
Formel:
C₉H₁₂O MW: 136,19 g/mol Kokpunkt: 109°C Smältpunkt: 60 °C Densitet: 0,99 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00002372 CAS nummer: 99-89-8 EINECS: 202-798-8 UN: 2430 ADR: 8,III |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 59.0 to 64.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...