2-Isopropylphenol ≥98.0%

Leverantör: TCI EUROPE
Fara

I0173-25G I0173-500G
TCIAI0173-25GEA 230 SEK
TCIAI0173-25G TCIAI0173-500G
2-Isopropylphenol ≥98.0%
2-Isopropylphenol
Formel: C₉H₁₂O
MW: 136,19 g/mol
Kokpunkt: 214°C
Smältpunkt: 14…16 °C
Densitet: 0,995 g/cm³ (20 °C)
Flampunkt: 104°C
Storage Temperature: Omgivande
MDL Number: MFCD00002224
CAS nummer: 88-69-7
EINECS: 201-852-8
UN: 3145
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4d11186c2a6f4e98bae336a74be5821bebc1719&vid=9f03b811a15a207cc21f9375f91e979013c484cf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR