4-Butylphenol ≥96.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Butyl-4-hydroxybenzene

B1026-25G B1026-5G
TCIAB1026-25GEA 1900 SEK
TCIAB1026-25G TCIAB1026-5G
4-Butylphenol ≥96.0%
4-Butylphenol
Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Kokpunkt: 245°C
Smältpunkt: 23…24 °C
Densitet: 0,9717 g/cm³ (20 °C)
Flampunkt: 121 °C
MDL Number: MFCD00041750
CAS nummer: 1638-22-8
EINECS: 216-672-5
UN: 2430
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c3c0836548a5a93be145d2eb9000d9aef4b769b&vid=5bdf90ba2ccb483e1a1e9546d85751adb3b05d2f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Brown clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR