2,4-Dinitrotoluen ≥99.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 1,3-Dinitro-4-methylbenzene
D0856-25G D0856-500G
TCIAD0856-25GEA 215 SEK
TCIAD0856-25G TCIAD0856-500G
2,4-Dinitrotoluen ≥99.0%
2,4-Dinitrotoluen
Formel: C₇H₆N₂O₄
MW: 182,14 g/mol
Kokpunkt: 300°C
Smältpunkt: 67…70 °C
Densitet: 1,379 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007172
CAS nummer: 121-14-2
EINECS: 204-450-0
UN: 3454
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74915c0e62624f0e3f8cc61a9af9b0b4978711a0&vid=181c0bc918f4cbc525517df6453b5e15160afd89&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 70.0 to 73.0 deg-C
Solubility in Methanol almost transparency