3-Methyl-2-nitroanisole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Methoxy-2-nitrotoluene, 1-Methoxy-3-methyl-2-nitrobenzene, Methyl 2-nitro-m-tolyl ether

M1196-25G M1196-5G
TCIAM1196-25GEA 1600 SEK
TCIAM1196-25G TCIAM1196-5G
3-Methyl-2-nitroanisole ≥98.0%
3-Methyl-2-nitroanisole
Formel: C₈H₉NO₃
MW: 167,16 g/mol
Kokpunkt: 170°C
Smältpunkt: 88…89 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007179
CAS nummer: 5345-42-6
EINECS: 226-295-8
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7d5ce7ad574e421cd50fb98d8a73658f8b1a088&vid=0d2a98a2c118f94d11fad0fb7fb1275b4640176f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 51.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR