2-Methyl-3-nitroanisole ≥97.0%

Leverantör: TCI
Varning

Synonyms: 1-Methoxy-2-methyl-3-nitrobenzene, Methyl 3-nitro-o-tolyl ether, 2-Methoxy-6-nitrotoluene

M1416-25G M1416-5G
TCIAM1416-25GEA 2925 SEK
TCIAM1416-25G TCIAM1416-5G
2-Methyl-3-nitroanisole ≥97.0%
2-Methyl-3-nitroanisole
Formel: C₈H₉NO₃
Kokpunkt: 94 - 96 °C (0,5 -rr)
Storage Temperature: Omgivande
MDL Number: MFCD00007159
CAS nummer: 4837-88-1
EINECS: 225-424-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=218c16d82e254a5b2481d01e3216e4072c3d5f9d&vid=04277ff3a1980470fdbf92ff1f2c9052cb2fb58a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR