4-Nitrotoluen ≥99.0%
Leverantör: TCI
|
Fara
|
N0276-25G
N0276-500G
TCIAN0276-25GEA
215
SEK
TCIAN0276-25G
TCIAN0276-500G
4-Nitrotoluen ≥99.0%
4-Nitrotoluen
Formel:
C₇H₇NO₂ MW: 137.14 g/mol Kokpunkt: 238°C Smältpunkt: 51 Flampunkt: 106 Storage Temperature: Omgivande |
MDL Number:
MFCD00007366 CAS nummer: 99-99-0 EINECS: 202-808-0 UN: 3446 ADR: 6.1 II Merck Index: 14,06650 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 51.0 to 55.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...