4-Hexylaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
H0890-25G H0890-5G
TCIAH0890-25GEA 2900 SEK
TCIAH0890-25G TCIAH0890-5G
4-Hexylaniline ≥98.0% (by GC, titration analysis)
4-Hexylaniline
Formel: C₁₂H₁₉N
MW: 177,29 g/mol
Kokpunkt: 285°C
Densitet: 0,91 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00007927
CAS nummer: 33228-45-4
EINECS: 251-409-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927a9f85db70cabbe1ebbd63d8883a190406b4d1&vid=d656dfd7ddc71e1863c49b702c28f2da60f61460&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %