4-Butylaniline ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Butylbenzenamine
B0272-25ML
B0272-500ML
B0272-100ML
TCIAB0272-25MLEA
245
SEK
TCIAB0272-25ML
TCIAB0272-500ML
TCIAB0272-100ML
4-Butylaniline ≥97.0% (by GC, titration analysis)
4-Butylaniline
Formel:
C₁₀H₁₅N MW: 149,24 g/mol Kokpunkt: 250°C Smältpunkt: –14 °C Densitet: 0,945 g/cm³ (20 °C) Flampunkt: 101°C |
MDL Number:
MFCD00007925 CAS nummer: 104-13-2 EINECS: 203-177-4 ADR: 6.1,III |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...