4-Isopropylaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: p-Cumidine, 4-Aminocumene
I0371-25ML I0371-500ML I0371-100ML
TCIAI0371-25MLEA 335 SEK
TCIAI0371-25ML TCIAI0371-500ML TCIAI0371-100ML
4-Isopropylaniline ≥98.0% (by GC, titration analysis)
4-Isopropylaniline
Formel: C₉H₁₃N
MW: 135,21 g/mol
Kokpunkt: 224°C
Smältpunkt: –63 °C
Densitet: 0,9526 g/cm³ (20 °C)
Flampunkt: 116°C
Storage Temperature: Omgivande
MDL Number: MFCD00007900
CAS nummer: 99-88-7
EINECS: 202-797-2
ADR: 8,III
Merck Index: 14,02620
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89bc49521ffbecdf544c4dd952110e1a4bd991a2&vid=859af7e2f6b06fd754b41010fb3a512162e5779b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %