4-sec-Butylaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 4-(sec-Butyl)aniline
B1256-25ML B1256-5ML
TCIAB1256-25MLEA 2425 SEK
TCIAB1256-25ML TCIAB1256-5ML
4-sec-Butylaniline ≥98.0% (by GC, titration analysis)
4-sec-Butylaniline
Formel: C₁₀H₁₅N
MW: 149,24 g/mol
Kokpunkt: 238°C
Densitet: 0,977 g/cm³ (20 °C)
Flampunkt: 107°C
MDL Number: MFCD00010223
CAS nummer: 30273-11-1
EINECS: 250-108-9
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=114e38302c007288b58a8f8e92ae431acfc22907&vid=114e38302c007288b58a8f8e92ae431acfc22907&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %