2-Propylaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
P1113-25G P1113-5G
TCIAP1113-25GEA 9600 SEK
TCIAP1113-25G TCIAP1113-5G
2-Propylaniline ≥98.0% (by GC, titration analysis)
2-Propylaniline
Formel: C₉H₁₃N
MW: 135,21 g/mol
Kokpunkt: 223°C
Densitet: 0,96 g/cm³ (20 °C)
Flampunkt: 98°C
MDL Number: MFCD00009803
CAS nummer: 1821-39-2
EINECS: 217-346-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8f6736d9d667861551a2423401b979643937117&vid=10635e7761a15dc41215f5d8e61f9c4c77b762be&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %