2-Ethylaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
E0060-25ML
E0060-500ML
TCIAE0060-25MLEA
215
SEK
TCIAE0060-25ML
TCIAE0060-500ML
2-Ethylaniline ≥98.0% (by GC, titration analysis)
2-Ethylaniline
Formel:
C₈H₁₁N MW: 121,18 g/mol Kokpunkt: 216°C Smältpunkt: –46 °C Densitet: 0,983 g/cm³ (20 °C) Flampunkt: 99°C Storage Temperature: Omgivande |
MDL Number:
MFCD00007751 CAS nummer: 578-54-1 EINECS: 209-424-2 UN: 2273 ADR: 6.1,III |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...