2-Iodo-4-nitrotoluene ≥97.0%
Leverantör: TCI
Varning
I0706-25G I0706-5G
TCIAI0706-25GEA 1350 SEK
TCIAI0706-25G TCIAI0706-5G
2-Iodo-4-nitrotoluene ≥97.0%
2-Iodo-4-nitrotoluene
Formel: C₇H₆INO₂
Kokpunkt: 165°C
MDL Number: MFCD00024329
CAS nummer: 7745-92-8
EINECS: 231-808-3
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0427c397ee7ae798fcdcc66dd51c1a584f68837&vid=6a8e0ed742d6aeca31e66123fad2fc2b8de2742f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 50.0 to 54.0 deg-C