4-Iodotoluene ≥99.0%

Leverantör: TCI
Varning

Synonyms: 1-Iodo-4-methylbenzene

I0218-25G I0218-500G I0218-100G
TCIAI0218-25GEA 455 SEK
TCIAI0218-25G TCIAI0218-500G TCIAI0218-100G
4-Iodotoluene ≥99.0%
4-Iodotoluene
Formel: C₇H₇I
Kokpunkt: 211,5 °C (1013 hPa)
Smältpunkt: 33…35 °C
Densitet: 1,68 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001059
CAS nummer: 624-31-7
EINECS: 210-841-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbaf4b78f4735e6c110e8c400a4167161f41e112&vid=9933bb546727ceb451bb694466c8b4d6f4159763&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR