4-Fluoro-2-nitrotoluene ≥96.0%

Leverantör: TCI EUROPE
Varning

F0218-25G F0218-5G
TCIAF0218-25GEA 790 SEK
TCIAF0218-25G TCIAF0218-5G
4-Fluoro-2-nitrotoluene ≥96.0%
4-Fluoro-2-nitrotoluene
Formel: C₇H₆FNO₂
MW: 155,13 g/mol
Kokpunkt: 139°C
Smältpunkt: 27 °C
Densitet: 1,2911 g/cm³ (0 °C)
Flampunkt: 98°C
MDL Number: MFCD00007200
CAS nummer: 446-10-6
EINECS: 207-161-8
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f01b8e6ff62e92987efd1c9d47b0fb76c31cd20b&vid=d11e715706cc322885912e502ebc8a6c2f8990ae&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR