5-Fluoro-2-nitrotoluene ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1-Fluoro-3-methyl-4-nitrobenzene

F0301-25G F0301-5G
TCIAF0301-25GEA 935 SEK
TCIAF0301-25G TCIAF0301-5G
5-Fluoro-2-nitrotoluene ≥98.0%
5-Fluoro-2-nitrotoluene
Formel: C₇H₆FNO₂
MW: 155.13 g/mol
Kokpunkt: 218
Smältpunkt: 28
Storage Temperature: Omgivande
MDL Number: MFCD00007283
CAS nummer: 446-33-3
EINECS: 207-164-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b932157ec53a433adcf10117bb40a1aac5d767c9&vid=f2d2edeb060b9a548cd3ada10389eef7d12ec9b2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR