2-Fluoro-6-nitrotoluene ≥98.0%

Leverantör: TCI EUROPE
Varning

F0696-25G F0696-5G
TCIAF0696-25GEA 1075 SEK
TCIAF0696-25G TCIAF0696-5G
2-Fluoro-6-nitrotoluene ≥98.0%
2-Fluoro-6-nitrotoluene
Formel: C₇H₆FNO₂
MW: 155,13 g/mol
Kokpunkt: 97°C
Smältpunkt: 7°C
Densitet: 1,2766 g/cm³ (20 °C)
Flampunkt: 89°C
MDL Number: MFCD00007197
CAS nummer: 769-10-8
EINECS: 212-203-3
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8506344e759f00c428a744dfcc7958c023ad373&vid=d54858a40fba1f58faeeb931c9955e80ed15faa4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR