4-Fluoro-2-iodotoluene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Fluoro-3-iodo-4-methylbenzene, 4-Fluoro-2-iodo-1-methylbenzene
F0613-25G
F0613-5G
TCIAF0613-25GEA
2125
SEK
TCIAF0613-25G
TCIAF0613-5G
4-Fluoro-2-iodotoluene ≥97.0%
4-Fluoro-2-iodotoluene
Formel:
C₇H₆FI MW: 236,03 g/mol Kokpunkt: 90°C Flampunkt: 87°C |
MDL Number:
MFCD00013709 CAS nummer: 13194-67-7 EINECS: 236-153-7 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...