3-Fluorotoluene ≥99.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1-Fluoro-3-methylbenzene
F0040-25ML
F0040-500ML
TCIAF0040-25MLEA
215
SEK
TCIAF0040-25ML
TCIAF0040-500ML
3-Fluorotoluene ≥99.0%
3-Fluorotoluene
Formel:
C₇H₇F MW: 110,13 g/mol Kokpunkt: 115°C Smältpunkt: –88 °C Densitet: 0,9986 g/cm³ (20 °C) Flampunkt: 9°C Storage Temperature: Omgivande |
MDL Number:
MFCD00000339 CAS nummer: 352-70-5 EINECS: 206-524-8 UN: 2388 ADR: 3,II Merck Index: 13,04207 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...