2-Fluorotoluene ≥99.0%

Leverantör: TCI
Fara

F0039-25ML F0039-500ML
TCIAF0039-25MLEA 215 SEK
TCIAF0039-25ML TCIAF0039-500ML
2-Fluorotoluene ≥99.0%
2-Fluorotoluene
Formel: C₇H₇F
Kokpunkt: 114°C
Smältpunkt: –62 °C
Densitet: 1,005 g/cm³ (20 °C)
Flampunkt: 12°C
Storage Temperature: Omgivande
MDL Number: MFCD00000322
CAS nummer: 95-52-3
EINECS: 202-428-5
UN: 2388
ADR: 3,II
Merck Index: 13,04207
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d63af630d71efceb84b67bad3655b9b6996fcf3c&vid=42f5e04bfd1b02d33c4a0f2de9517a31fb34056f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR