α,α,α,2-Tetrafluorotoluene ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Fluoro-2-(trifluoromethyl)benzene, 2-Fluorobenzotrifluoride

F0389-25G F0389-5G
TCIAF0389-25GEA 550 SEK
TCIAF0389-25G TCIAF0389-5G
α,α,α,2-Tetrafluorotoluene ≥98.0%
α,α,α,2-Tetrafluorotoluene
Formel: C₇H₄F₄
MW: 164,1 g/mol
Kokpunkt: 116°C
Smältpunkt: –51,15 °C
Densitet: 1,293 g/cm³ (26 °C)
Flampunkt: 17°C
MDL Number: MFCD00000374
CAS nummer: 392-85-8
EINECS: 206-880-4
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b80c03a6f34fa5f955f2c18c78e4751370fae4b&vid=54eb1380bfee5b5a41619c0c17c5eac48c29c38c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR