3-Chloro-4-nitrotoluene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Chloro-4-methyl-1-nitrobenzene

C2277-25G C2277-5G
TCIAC2277-25GEA 1225 SEK
TCIAC2277-25G TCIAC2277-5G
3-Chloro-4-nitrotoluene ≥98.0%
3-Chloro-4-nitrotoluene
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Kokpunkt: 101°C
Smältpunkt: 24 °C
Densitet: 1,324 g/cm³ (20 °C)
MDL Number: MFCD02683043
CAS nummer: 38939-88-7
EINECS: 254-199-6
UN: 3457
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea1476b4ce9f52870c8c0ba254894527bd134f70&vid=7816835d623c8c8f45b9833a7f5391bcaf98ef5c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR