2-Chloro-5-nitrotoluene ≥98.0%
Leverantör: TCI
Varning
Synonyms: 1-Chloro-2-methyl-4-nitrobenzene
C1233-25G C1233-500G
TCIAC1233-25GEA 510 SEK
TCIAC1233-25G TCIAC1233-500G
2-Chloro-5-nitrotoluene ≥98.0%
2-Chloro-5-nitrotoluene
Formel: C₇H₆ClNO₂
MW: 171.58 g/mol
Kokpunkt: 96°C
Storage Temperature: Omgivande
MDL Number: MFCD00041430
CAS nummer: 13290-74-9
EINECS: 236-306-8
UN: 3457
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3ada871ca903aded6a0504232428efb60a97fce&vid=d861f2b8950c005a9329d6fb4d13817e068cbfb9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 45.0 deg-C
Solubility in Methanol almost transparency