2-Nitrobenzyl chloride ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: 1-(Chloromethyl)-2-nitrobenzene, 2-Nitrobenzyl chloride, α-Chloro-o-nitrotoluene, alpha-Chloro-2-nitrotoluene
N0182-25G N0182-5G
TCIAN0182-25GEA 1375 SEK
TCIAN0182-25G TCIAN0182-5G
2-Nitrobenzyl chloride ≥98.0% (by GC, titration analysis)
2-Nitrobenzyl chloride
Formel: C₇H₆ClNO₂
MW: 171.58 g/mol
Kokpunkt: 133°C
Storage Temperature: Omgivande
MDL Number: MFCD00007185
CAS nummer: 612-23-7
EINECS: 210-300-5
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f70b98add4808e786715783cce9e32742e87d58&vid=ad07d2360068ed9016303a3dd85db567cbc33825&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 47.0 to 50.0 deg-C