4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥98.0%

Leverantör: TCI
Fara

Synonyms: 1-Chloro-2,6-dinitro-4-(trifluoromethyl)benzene, 4-Chloro-3,5-dinitrobenzotrifluoride

C0994-10G C0994-25G
TCIAC0994-10GEA 565 SEK
TCIAC0994-10G TCIAC0994-25G
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥98.0%
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Formel: C₇H₂ClF₃N₂O₄
Kokpunkt: 250 °C (1013 hPa)
Smältpunkt: 56…58 °C
Densitet: 1,6 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007068
CAS nummer: 393-75-9
EINECS: 206-889-3
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36ef8fe724b1e0ad59c669772847999834b3c7c2&vid=d3df48ac7abff6e1370e763b1e7c6c9629098d94&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 58.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR