2-Chloro-4-fluorotoluene ≥98.0%

Leverantör: TCI
Varning

C1227-25G C1227-500G
TCIAC1227-25GEA 270 SEK
TCIAC1227-25G TCIAC1227-500G
2-Chloro-4-fluorotoluene ≥98.0%
2-Chloro-4-fluorotoluene
Formel: C₇H₆ClF
Kokpunkt: 153°C
Densitet: 1,1972 g/cm³ (20 °C)
Flampunkt: 50°C
Storage Temperature: Omgivande
MDL Number: MFCD00000572
CAS nummer: 452-73-3
EINECS: 207-209-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6180a8e1a82536ff7fa5e899dce998d537af191d&vid=305421020c19226e472d1cc3ac8bdbeb2bbfcfcf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR