(4-Chlorophenyl)acetone ≥97.0%
Leverantör: TCI
Synonyms:
4-Chlorophenylacetone
C2526-25G
C2526-5G
TCIAC2526-25GEA
2125
SEK
TCIAC2526-25G
TCIAC2526-5G
(4-Chlorophenyl)acetone ≥97.0%
(4-Chlorophenyl)acetone
Formel:
C₉H₉ClO Kokpunkt: 216°C |
MDL Number:
MFCD00045214 CAS nummer: 5586-88-9 EINECS: 226-986-4 |
|
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...