α,4-Dichlorotoluene ≥98.0%
Leverantör: TCI
Fara
D0421-25G D0421-500G
TCIAD0421-25GEA 470 SEK
TCIAD0421-25G TCIAD0421-500G
α,4-Dichlorotoluene ≥98.0%
α,4-Dichlorotoluene
Formel: C₇H₆Cl₂
Kokpunkt: 214 °C (1013 hPa)
Smältpunkt: 28…30 °C
Densitet: 1,259 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000914
CAS nummer: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0c9e39d248d340d96f9f6c34e26ba236bb7b3a2&vid=ebe0f78b81169f82b40bd9b3534c54655368c7cd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency