α,α,α,2-Tetraklortoluen ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Klorbenzotriklorid
C0586-25G C0586-500G
TCIAC0586-25GEA 215 SEK
TCIAC0586-25G TCIAC0586-500G
α,α,α,2-Tetraklortoluen ≥98.0%
α,α,α,2-Tetraklortoluen
Formel: C₇H₄Cl₄
MW: 229,92 g/mol
Kokpunkt: 264 °C (1013 hPa)
Smältpunkt: 29…31 °C
Densitet: 1,5187 g/cm³ (20 °C)
MDL Number: MFCD00000788
CAS nummer: 2136-89-2
EINECS: 218-377-7
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06680b9468725b74ab93b55621a3bea3b1296730&vid=6f89116be2a8f9a71ceba6afaea41b4b85614029&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %