6-Hydroxy-m-toluic acid ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 5-Methylsalicylic acid, 2,5-Cresotic acid
C0410-25G C0410-5G
TCIAC0410-25GEA 1325 SEK
TCIAC0410-25G TCIAC0410-5G
6-Hydroxy-m-toluic acid ≥98.0%
6-Hydroxy-m-toluic acid
Formel: C₈H₈O₃
MW: 152,15 g/mol
Kokpunkt: 313 °C (1013 hPa)
Smältpunkt: 152 °C
Densitet: 1,31 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002461
CAS nummer: 89-56-5
EINECS: 201-918-6
Merck Index: 14,02582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c9b1f226d08a55ab7f8ab51768a0fee694d8eeb&vid=c07eb07cfdc35b86428ee0b4bca6151083e16f88&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 153.0 deg-C
Solubility in Methanol very faint turbidity