6-Nitro-o-toluic acid ≥98.0%

Leverantör: TCI
Varning

M1002-10G M1002-25G
TCIAM1002-10GEA 575 SEK
TCIAM1002-10G TCIAM1002-25G
6-Nitro-o-toluic acid ≥98.0%
6-Nitro-o-toluic acid
Formel: C₈H₇NO₄
Kokpunkt: 353 °C (1013 hPa)
Smältpunkt: 151…153 °C
MDL Number: MFCD00007267
CAS nummer: 13506-76-8
EINECS: 236-833-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57c251c816d3900ca82f302cd32a95634579a9d1&vid=3d442566f83e9620ffcedd0c80e9a9af2238601e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 158.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR