5-Nitro-o-toluic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Methyl-5-nitrobenzoic acid
M1716-1G M1716-5G
TCIAM1716-1GEA 230 SEK
TCIAM1716-1G TCIAM1716-5G
5-Nitro-o-toluic acid ≥98.0% (by GC, titration analysis)
5-Nitro-o-toluic acid
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Smältpunkt: 179…181 °C
Densitet: 1,49 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007371
CAS nummer: 1975-52-6
EINECS: 217-829-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15d104c4f3ed1899ec91625f8219880cfa1a1357&vid=94ba9a73790e26aff63ea8fd17ba3195a23e20b0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 177.0 to 181.0 deg-C