Propiophenone ≥98.0%
Leverantör: TCI
Fara
Synonyms: Phenyl ethyl ketone, Ethyl phenyl ketone, 1-Phenyl-1-propanone
P0519-25G P0519-500G
TCIAP0519-25GEA 430 SEK
TCIAP0519-25G TCIAP0519-500G
Propiophenone ≥98.0%
Propiophenone
Formel: C₆H₅C(O)CH₂CH₃
Kokpunkt: 216°C
Smältpunkt: 18 °C
Densitet: 1,01 g/cm³ (20 °C)
Flampunkt: 92°C
Storage Temperature: Omgivande
MDL Number: MFCD00009309
CAS nummer: 93-55-0
EINECS: 202-257-6
Merck Index: 14,07830
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e34c7b8eaf3ea974e70e8eace2c8b9aec54ad16&vid=6b115d9126ef33a29e0f649597246f00db797dad&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %