Butyrophenone ≥99.0%
Leverantör: TCI EUROPE
Synonyms: 1-Phenylbutan-1-one, Phenyl propyl ketone, 1-Phenyl-1-butanone
B0769-25ML B0769-500ML B0769-100ML
TCIAB0769-25MLEA 255 SEK
TCIAB0769-25ML TCIAB0769-500ML TCIAB0769-100ML
Butyrophenone ≥99.0%
Butyrophenone
Formel: C₆H₅C(O)(CH₂)₂CH₃
MW: 148,2 g/mol
Kokpunkt: 220 °C (1013 hPa)
Densitet: 0,988 g/cm³ (20 °C)
Flampunkt: 88°C
Storage Temperature: Omgivande
MDL Number: MFCD00009397
CAS nummer: 495-40-9
EINECS: 207-799-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=480a64492a3195f352d5b0f17d550c62fc4db434&vid=2909b9c529d4e49a9bfc9def1a1dacb2cb6000d7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %