4-tert-Butylbenzonitrile ≥98.0%
Leverantör: TCI
|
Fara
|
B1576-25ML
B1576-5ML
TCIAB1576-25MLEA
2125
SEK
TCIAB1576-25ML
TCIAB1576-5ML
4-tert-Butylbenzonitrile ≥98.0%
4-tert-Butylbenzonitrile
Formel:
C₁₁H₁₃N Kokpunkt: 112°C Densitet: 0,9492 g/cm³ (20 °C) |
MDL Number:
MFCD00075840 CAS nummer: 4210-32-6 EINECS: 224-137-2 UN: 3276 ADR: 6.1,III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...