4-Isopropylbenzaldehyde ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
Cuminaldehyde
I0168-25G
I0168-500G
TCIAI0168-25GEA
285
SEK
TCIAI0168-25G
TCIAI0168-500G
4-Isopropylbenzaldehyde ≥97.0%
4-Isopropylbenzaldehyde
Formel:
C₁₀H₁₂O MW: 148,2 g/mol |
MDL Number:
MFCD00006953 CAS nummer: 122-03-2 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |