4-Bromo-2-nitrotoluene ≥98.0%
Leverantör: TCI EUROPE
Varning
B1843-25G B1843-5G
TCIAB1843-25GEA 1125 SEK
TCIAB1843-25G TCIAB1843-5G
4-Bromo-2-nitrotoluene ≥98.0%
4-Bromo-2-nitrotoluene
Formel: C₇H₆BrNO₂
MW: 216,03 g/mol
Kokpunkt: 256°C
Smältpunkt: 43…46 °C
MDL Number: MFCD00041243
CAS nummer: 60956-26-5
EINECS: 262-536-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83ecf00f9d0fef08a78228ededf159b02955309a&vid=a4eaed77561f84491b2a2e7b28ced9e52073b972&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 48.0 deg-C