2-Bromo-5-nitrotoluene ≥98.0%
Leverantör: TCI
Varning
Synonyms: 1-Bromo-2-methyl-4-nitrobenzene
B1853-25G B1853-5G
TCIAB1853-25GEA 2975 SEK
TCIAB1853-25G TCIAB1853-5G
2-Bromo-5-nitrotoluene ≥98.0%
2-Bromo-5-nitrotoluene
Formel: C₇H₆BrNO₂
MW: 216.03 g/mol
Kokpunkt: 143°C
Smältpunkt: 78
MDL Number: MFCD00007281
CAS nummer: 7149-70-4
EINECS: 230-481-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=114528b994d9a94e06d61382d9123ef41b8e2898&vid=fff409429bbbd1d6efbed53371d99f4736ba8663&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 80.0 deg-C