4-Bromo-2-fluorotoluene ≥97.0%

Leverantör: TCI
Varning

Synonyms: 1-Bromo-3-fluoro-4-methylbenzene

B1716-25G B1716-5G
TCIAB1716-25GEA 830 SEK
TCIAB1716-25G TCIAB1716-5G
4-Bromo-2-fluorotoluene ≥97.0%
4-Bromo-2-fluorotoluene
Formel: C₇H₆BrF
Kokpunkt: 100°C
Densitet: 1,5 g/cm³ (20 °C)
Flampunkt: 76°C
MDL Number: MFCD00013551
CAS nummer: 51436-99-8
EINECS: 257-201-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0009b5a9c1ccf5da7f1176705a6b8a3d1fe16def&vid=e28d906fccb287e21f05ef3ec354d48e1251073d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR