α-Bromo-3-fluorotoulene ≥97.0%
Leverantör: TCI EUROPE
Fara
Synonyms: α-Bromo-3-fluorotoluene, α-Bromo-m-fluorotoulene, alpha-Bromo-m-fluorotoulene, 1-(Bromomethyl)-3-fluorobenzene, m-Fluorobenzyl bromide, 3-Fluorobenzyl bromide
F0208-25G F0208-5G
TCIAF0208-25GEA 735 SEK
TCIAF0208-25G TCIAF0208-5G
α-Bromo-3-fluorotoulene ≥97.0%
α-Bromo-3-fluorotoulene
Formel: C₇H₆BrF
MW: 189,03 g/mol
Kokpunkt: 200°C
Densitet: 1,541 g/cm³ (20 °C)
Flampunkt: 61°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000340
CAS nummer: 456-41-7
EINECS: 207-263-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdf8df0602f1656bf49690f3b888f165174ab88c&vid=10020dfe9d1414ce169633dac7b75d4811cbdbde&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %