α-Bromo-2,3,5,6-tetrafluorotoluene ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2,3,5,6-Tetrafluorobenzyl bromide, 2,3,5,6-Tetrafluorobenzylbromide

T1886-1G T1886-5G
TCIAT1886-1GEA 1125 SEK
TCIAT1886-1G TCIAT1886-5G
α-Bromo-2,3,5,6-tetrafluorotoluene ≥97.0%
α-Bromo-2,3,5,6-tetrafluorotoluene
Formel: C₇H₃BrF₄
MW: 243 g/mol
Kokpunkt: 80°C
MDL Number: MFCD00013549
CAS nummer: 53001-73-3
EINECS: 258-289-6
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e809decf6b89aff04991c5b447c4df583fb6f31&vid=7493456eee2ea8aab734ba29155100793e296e11&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR