Pentafluorobenzyl bromide ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide
P0809-1G P0809-25G P0809-5G
TCIAP0809-1GEA 215 SEK
TCIAP0809-1G TCIAP0809-25G TCIAP0809-5G
Pentafluorobenzyl bromide ≥98.0%
Pentafluorobenzyl bromide
Formel: (C₆F₅)CH₂Br
MW: 260,99 g/mol
Kokpunkt: 176°C
Densitet: 1,72 g/cm³ (20 °C)
Flampunkt: 83°C
Storage Temperature: Omgivande
MDL Number: MFCD00000299
CAS nummer: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46a8d8990d9350210c7eecdad657f210818e80be&vid=2f6c65e6cb425790ce46264d155ba84c753ec39b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %