α-Bromo-4-chlorotoulene ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide
B0576-25G
B0576-5G
TCIAB0576-25GEA
1300
SEK
TCIAB0576-25G
TCIAB0576-5G
α-Bromo-4-chlorotoulene ≥98.0%
α-Bromo-4-chlorotoulene
Formel:
C₇H₆BrCl MW: 205.48 g/mol Kokpunkt: 110°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00040714 CAS nummer: 622-95-7 EINECS: 210-760-7 UN: 3261 ADR: 8 II |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...