α,3-Dibromotoluene ≥98.0%

Leverantör: TCI
Fara

Synonyms: 3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene

B1570-25G B1570-5G
TCIAB1570-25GEA 845 SEK
TCIAB1570-25G TCIAB1570-5G
α,3-Dibromotoluene ≥98.0%
α,3-Dibromotoluene
Formel: C₇H₆Br₂
Kokpunkt: 135°C
Smältpunkt: 41…44 °C
Densitet: 1,56 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00000176
CAS nummer: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc048cf519e13416e7c7ef445132493260363d10&vid=8fb90de53fb1961f3525cf33b9cc52501f402cfb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 44.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR