4-Hydroxyphenylboronic acid pinacol ester ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Hydroxybenzeneboronic acid pinacol ester
T1954-1G
T1954-5G
TCIAT1954-1GEA
215
SEK
TCIAT1954-1G
TCIAT1954-5G
4-Hydroxyphenylboronic acid pinacol ester ≥98.0%
4-Hydroxyphenylboronic acid pinacol ester
Formel:
C₁₂H₁₇BO₃ MW: 220,08 g/mol |
MDL Number:
MFCD02093756 CAS nummer: 269409-70-3 |
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...