3,5-Dibromotoluene ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1,3-Dibromo-5-methylbenzene
D3376-25G D3376-5G
TCIAD3376-25GEA 1450 SEK
TCIAD3376-25G TCIAD3376-5G
3,5-Dibromotoluene ≥98.0%
3,5-Dibromotoluene
Formel: C₇H₆Br₂
MW: 249,93 g/mol
Kokpunkt: 246°C
MDL Number: MFCD00013528
CAS nummer: 1611-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d34fe65afda93372d4eab09ff27c106bcfbb30&vid=46763448cf6702a54364cbcea777c61b34902079&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C